Orders LinkContactUsLinkComponent
SkillPak™ BIO Pre-Packed Columns
Supplier:   TOSOH Bioscience
SkillPak™ BIO Pre-Packed Columns
Supplier:  TOSOH Bioscience
Certificates
SkillPak™ BIO Pre-Packed Columns
SkillPak™ BIO Pre-Packed Columns
51 Options Available Below
Product Details & Documents
SkillPak™ BIO pre-packed columns are designed for a fast method development and scale-up of multi-column chromatography (MCC) or continuous chromatography processes. They are pre-packed with TOYOPEARL® process chromatography media for bioseparations.
  • MCC-dedicated dimensions and packing procedures
  • Comprehensive resins for MCC applications
  • Multi-column kits with single resin lot and low column-to-column variability
They are ready to use and show excellent physical strength and ideal pressure-flow characteristic for downstrem processing in multi-column (MCC) or continuous (CC) operations. Pre-packed with TOYOPEARL®process chromatography media, these columns offer superior separation of biomolecules, such as monoclonal antibodies. SkillPak™ BIO columns are available in multi-column kits packed with a single resin lot and low column-to-column variability to ensure low comparability between columns in MCC and CC.
All SkillPak BIO columns guarantee optimal performance and can be operated on all MCC and CC systems, as well as standard low-or medium-pressure liquid chromatography systems. They are reproducibly packed and take into account the varying compressibility of each resin.
Product Family Options
1 ml columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Capacity
    Packing material
    Availability
    Price
  • Method development kit
    Protein A
    PP
    1000 Å
    45 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 2 630,00 €
    2 630,00 €
    1 * 6 ST
       2 630,00 €
  • Method development kit
    Protein A
    PP
    1000 Å
    45 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 3 680,00 €
    3 680,00 €
    1 * 10 ST
       3 680,00 €
  • Method development kit
    Protein L
    PP
    1000 Å
    45 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 2 630,00 €
    2 630,00 €
    1 * 6 ST
       2 630,00 €
  • Method development kit
    Protein L
    PP
    1000 Å
    45 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 3 680,00 €
    3 680,00 €
    1 * 10 ST
       3 680,00 €
  • Method development kit
    Sulfate
    PP
    1000 Å
    45 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 1 750,00 €
    1 750,00 €
    1 * 4 ST
       1 750,00 €
  • Method development kit
    Proprietary
    PP
    >1000 Å
    45 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 1 750,00 €
    1 750,00 €
    1 * 4 ST
       1 750,00 €
  • Method development kit
    Sulfopropyl
    PP
    1000 Å
    75 µm
    8,10 mm
    20 mm
    1 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 1 750,00 €
    1 750,00 €
    1 * 4 ST
       1 750,00 €
5 ml columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Capacity
    Packing material
    Availability
    Price
  • Method development kit
    Quarternary amine
    PP
    >1000 Å
    35 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL SuperQ-650S
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 3 590,00 €
    3 590,00 €
    1 * 4 ST
       3 590,00 €
  • Method development kit
    Protein A
    PP
    1000 Å
    45 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 5 940,00 €
    5 940,00 €
    1 * 6 ST
       5 940,00 €
  • Method development kit
    Protein A
    PP
    1000 Å
    45 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 8 970,00 €
    8 970,00 €
    1 * 10 ST
       8 970,00 €
  • Method development kit
    Protein L
    PP
    1000 Å
    45 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 5 940,00 €
    5 940,00 €
    1 * 6 ST
       5 940,00 €
  • Method development kit
    Protein L
    PP
    1000 Å
    45 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 8 970,00 €
    8 970,00 €
    1 * 10 ST
       8 970,00 €
  • Method development kit
    Sulfate
    PP
    1000 Å
    45 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 3 590,00 €
    3 590,00 €
    1 * 4 ST
       3 590,00 €
  • Method development kit
    Proprietary
    PP
    >1000 Å
    45 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 3 590,00 €
    3 590,00 €
    1 * 4 ST
       3 590,00 €
  • Method development kit
    Sulfopropyl
    PP
    1000 Å
    75 µm
    16,00 mm
    25 mm
    5 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 3 590,00 €
    3 590,00 €
    1 * 4 ST
       3 590,00 €
10 ml columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Capacity
    Packing material
    Availability
    Price
  • Method Validation Kit
    Quarternary amine
    PP
    1000 Å
    35 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL SuperQ-650S
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 4 510,00 €
    4 510,00 €
    1 * 4 ST
       4 510,00 €
  • Method Validation Kit
    Protein A
    PP
    1000 Å
    45 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 7 590,00 €
    7 590,00 €
    1 * 6 ST
       7 590,00 €
  • Method Validation Kit
    Protein A
    PP
    1000 Å
    45 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 11 270,00 €
    11 270,00 €
    1 * 10 ST
       11 270,00 €
  • Method Validation Kit
    Protein L
    PP
    1000 Å
    45 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 7 590,00 €
    7 590,00 €
    1 * 6 ST
       7 590,00 €
  • Method Validation Kit
    Protein L
    PP
    1000 Å
    45 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 11 270,00 €
    11 270,00 €
    1 * 10 ST
       11 270,00 €
  • Method Validation Kit
    Sulfate
    PP
    1000 Å
    45 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 4 510,00 €
    4 510,00 €
    1 * 4 ST
       4 510,00 €
  • Method Validation Kit
    Proprietary
    PP
    >1000 Å
    45 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 4 510,00 €
    4 510,00 €
    1 * 4 ST
       4 510,00 €
  • Method Validation Kit
    Sulfopropyl
    PP
    1000 Å
    75 µm
    16,00 mm
    51 mm
    10 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 4 510,00 €
    4 510,00 €
    1 * 4 ST
       4 510,00 €
25 ml columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Capacity
    Packing material
    Availability
    Price
  • Method Validation Kit
    Quarternary amine
    PP
    1000 Å
    35 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL SuperQ-650S
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 200,00 €
    3 200,00 €
    1 * 1 ST
       3 200,00 €
  • Method Validation Kit
    Quarternary amine
    PP
    1000 Å
    35 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL SuperQ-650S
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 10 030,00 €
    10 030,00 €
    1 * 4 ST
       10 030,00 €
  • Method Validation Kit
    Protein A
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 660,00 €
    3 660,00 €
    1 * 1 ST
       3 660,00 €
  • Method Validation Kit
    Protein A
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 17 810,00 €
    17 810,00 €
    1 * 6 ST
       17 810,00 €
  • Method Validation Kit
    Protein A
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 25 070,00 €
    25 070,00 €
    1 * 10 ST
       25 070,00 €
  • Method Validation Kit
    Protein L
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 660,00 €
    3 660,00 €
    1 * 1 ST
       3 660,00 €
  • Method Validation Kit
    Protein L
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 17 810,00 €
    17 810,00 €
    1 * 6 ST
       17 810,00 €
  • Method Validation Kit
    Protein L
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 25 070,00 €
    25 070,00 €
    1 * 10 ST
       25 070,00 €
  • Method Validation Kit
    Sulfate
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 200,00 €
    3 200,00 €
    1 * 1 ST
       3 200,00 €
  • Method Validation Kit
    Sulfate
    PP
    1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 10 030,00 €
    10 030,00 €
    1 * 4 ST
       10 030,00 €
  • Method Validation Kit
    Proprietary
    PP
    >1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 200,00 €
    3 200,00 €
    1 * 1 ST
       3 200,00 €
  • Method Validation Kit
    Proprietary
    PP
    >1000 Å
    45 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 10 030,00 €
    10 030,00 €
    1 * 4 ST
       10 030,00 €
  • Method Validation Kit
    Sulfopropyl
    PP
    1000 Å
    75 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 200,00 €
    3 200,00 €
    1 * 1 ST
       3 200,00 €
  • Method Validation Kit
    Sulfopropyl
    PP
    1000 Å
    75 µm
    25,00 mm
    51 mm
    25 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 10 030,00 €
    10 030,00 €
    1 * 4 ST
       10 030,00 €
100 ml columns
Product Information
  • Column Format
    Phase
    Column hardware
    Pore size
    Particle Size
    Ø int.
    Length
    Capacity
    Packing material
    Availability
    Price
  • Semi-preparative
    Quarternary amine
    PP
    1000 Å
    35 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL SuperQ-650S
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 290,00 €
    5 290,00 €
    1 * 1 ST
       5 290,00 €
  • Semi-preparative
    Quarternary amine
    PP
    1000 Å
    35 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL SuperQ-650S
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 16 930,00 €
    16 930,00 €
    1 * 4 ST
       16 930,00 €
  • Semi-preparative
    Protein A
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 880,00 €
    6 880,00 €
    1 * 1 ST
       6 880,00 €
  • Semi-preparative
    Protein A
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 34 370,00 €
    34 370,00 €
    1 * 6 ST
       34 370,00 €
  • Semi-preparative
    Protein A
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL AF-rProtein A HC-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52 670,00 €
    52 670,00 €
    1 * 10 ST
       52 670,00 €
  • Semi-preparative
    Protein L
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 880,00 €
    6 880,00 €
    1 * 1 ST
       6 880,00 €
  • Semi-preparative
    Protein L
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 34 370,00 €
    34 370,00 €
    1 * 6 ST
       34 370,00 €
  • Semi-preparative
    Protein L
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL AF-rProtein L-650F
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52 670,00 €
    52 670,00 €
    1 * 10 ST
       52 670,00 €
  • Semi-preparative
    Sulfate
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 290,00 €
    5 290,00 €
    1 * 1 ST
       5 290,00 €
  • Semi-preparative
    Sulfate
    PP
    1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL Sulfate-650F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 16 930,00 €
    16 930,00 €
    1 * 4 ST
       16 930,00 €
  • Semi-preparative
    Proprietary
    PP
    >1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 290,00 €
    5 290,00 €
    1 * 1 ST
       5 290,00 €
  • Semi-preparative
    Proprietary
    PP
    >1000 Å
    45 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL NH2-750F
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 16 930,00 €
    16 930,00 €
    1 * 4 ST
       16 930,00 €
  • Semi-preparative
    Sulfopropyl
    PP
    1000 Å
    75 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 290,00 €
    5 290,00 €
    1 * 1 ST
       5 290,00 €
  • Semi-preparative
    Sulfopropyl
    PP
    1000 Å
    75 µm
    50,00 mm
    51 mm
    100 ml
    TOYOPEARL GigaCap S-650M
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 16 930,00 €
    16 930,00 €
    1 * 4 ST
       16 930,00 €