Orders LinkContactUsLinkComponent
Avantor® Rigid Outer Support Containers
Avantor® Rigid Outer Support Containers
Supplier:   VWR Collection
Avantor® Rigid Outer Support Containers
Supplier:  VWR Collection
Certificates
Avantor® Rigid Outer Support Containers
Avantor® Rigid Outer Support Containers
12 Options Available Below
Product Details & Documents
3D cubic bags can fit in existing support containers. These support containers are available in either High Density Polyethylene (HDPE) and Polypropelene (PP), or stainless steel material.
  • Standard 3D cubic bags are available in 100, 200, and 1000 L volumes with 3 top ports
  • Standard 3D cubic bags are available with or without a bottom port
  • Cleanroom and shipping variants are available in either material format
  • Plastic containers can collapse for easy storage
  • Stainless steel containers are ISTA certified for shipping
Avantor® 3D cubic bags are made from a proprietary film which features a polyethelene inner and outer layer with gas and moisture barrier layer between. The film has high capacity and flexibility, and is resistant to a wide range of chemicals.
Certifications: Stainless steel containers meet ISTA 3,1B certification.
Product Family Options
Plastic, foldable, cleanroom version
Product Information
  • Capacity
    Material
    Availability
    Price
  • 734-3421
    Rigid outer support container, foldable, cleanroom version
    Rigid outer support container, foldable, cleanroom version
    250 L
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 630,00 €
    1 630,00 €
    1 * 1 ST
       1 630,00 €
  • 734-3422
    Rigid outer support container, foldable, cleanroom version
    Rigid outer support container, foldable, cleanroom version
    500 L
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 830,00 €
    3 830,00 €
    1 * 1 ST
       3 830,00 €
  • 734-3423
    Rigid outer support container, foldable, cleanroom version
    Rigid outer support container, foldable, cleanroom version
    1000 L
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 070,00 €
    4 070,00 €
    1 * 1 ST
       4 070,00 €
Plastic, foldable, transport version
Product Information
  • Capacity
    Material
    Availability
    Price
  • 734-3424
    Rigid outer support container, foldable, transport version
    Rigid outer support container, foldable, transport version
    250 L
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 050,00 €
    2 050,00 €
    1 * 1 ST
       2 050,00 €
  • 734-3425
    Rigid outer support container, foldable, transport version
    Rigid outer support container, foldable, transport version
    500 L
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 970,00 €
    3 970,00 €
    1 * 1 ST
       3 970,00 €
  • 734-3426
    Rigid outer support container, foldable, transport version
    Rigid outer support container, foldable, transport version
    1000 L
    Plastic
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 220,00 €
    4 220,00 €
    1 * 1 ST
       4 220,00 €
Stainless steel, cleanroom version
Product Information
  • Capacity
    Material
    Availability
    Price
  • 734-3430
    Rigid outer support container, cleanroom version
    Rigid outer support container, cleanroom version
    200 L
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 340,00 €
    4 340,00 €
    1 * 1 ST
       4 340,00 €
  • 734-3431
    Rigid outer support container, cleanroom version
    Rigid outer support container, cleanroom version
    500 L
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 400,00 €
    6 400,00 €
    1 * 1 ST
       6 400,00 €
  • 734-3432
    Rigid outer support container, cleanroom version
    Rigid outer support container, cleanroom version
    1000 L
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9 300,00 €
    9 300,00 €
    1 * 1 ST
       9 300,00 €
Stainless steel, transport version
Product Information
  • Capacity
    Material
    Availability
    Price
  • 734-3427
    Rigid outer support container, transport version
    Rigid outer support container, transport version
    200 L
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12 230,00 €
    12 230,00 €
    1 * 1 ST
       12 230,00 €
  • 734-3428
    Rigid outer support container, transport version
    Rigid outer support container, transport version
    500 L
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14 960,00 €
    14 960,00 €
    1 * 1 ST
       14 960,00 €
  • 734-3429
    Rigid outer support container, transport version
    Rigid outer support container, transport version
    1000 L
    Stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17 190,00 €
    17 190,00 €
    1 * 1 ST
       17 190,00 €
Complementary Products
Product Information
  • Catalog #
    Description
    Capacity
    Availability
    Price
  • 734-2961
    3D cubic bags with 2× 50 cm silicone platinum ports
    Port 1: 1,27×1,90 mm, with pinch clamp and 1× male MPX
    Port 2: 1,27×1,90 mm, with pinch clamp and 1× female MPX
    500 l
    Log in for availability
    1 * 1 ST - 565,00 €
  • 734-2960
    3D cubic bags with 2× 50 cm silicone platinum ports
    Port 1: 1,27×1,90 mm, with pinch clamp and 1× male MPX
    Port 2: 1,27×1,90 mm, with pinch clamp and 1× female MPX
    200 l
    Log in for availability
    1 * 5 ST - 4 120,00 €