ContactUsLinkComponent
Accessries for Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Accessries for Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Catalog # COCO08467-64
Supplier:  CONTROL
undefined
Accessries for Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Catalog # COCO08467-64
Supplier:  CONTROL
Specifications
  • Description:
    Type-K high-temperature ceramic fiber-insulated probe mini-connector, exposed 152,4 cm (5'), 20-gauge (60" cable length), for type-K mini-connector thermocouple meter, range: –150...1093 °C, accuracy: ±1,1 °C (3,0 °F) or ±0,4% reading above 100 °C (212 °F); ±2,2 °C (3,6 °F) or 2% of reading between –50...99 °C (–58...211 °F); ±4 °C (7,2 °F) or ±4% of reading between –51...–150 °C (–60...–238 °F), whichever is greater
  • Pk:
    1