ContactUsLinkComponent
Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Catalog # COCO20250-20
Supplier:  CONTROL
undefined
Digi-Sense® Pro 4-Input Data Logging Thermocouple Thermometer
Catalog # COCO20250-20
Supplier:  CONTROL
Specifications
  • Range:
    −200...1372 °C
  • Resolution:
    0,1 °C; 1 °C (above 999,9 °C)
  • Data memory - No. of readings:
    10000 data points
  • Description:
    Pro 4-input data logging thermocouple thermometer, Type K, NIST-traceable calibration, 4 probe channels
  • Output:
    USB
  • Batteries:
    9 V battery
  • Display:
    Backlit LCD
  • Accuracy:
    ±0,15% of reading 1,0 °C, ±0,5% of reading 2,0 °C below –100 °C (±0,15% of reading 1,8 °F, ±0,5% of reading 3,6 °F below –148 °F)
  • Pk:
    1
Specifications
About this item
Rugged, heavy-duty construction protects against knocks and jars.
  • Reliable measurements, guaranteed!
  • Precalibrated to NIST-traceable standards
  • Relative time stamp provides a time reference for major events
  • Electronic off set function allows compensation of probe errors
  • Infrared input port allows safe, noncontact measurements with optional IR wand
These professional type K thermometers provide accurate and reliable temperature measurement in an ergonomic design. Double-molded heavy-duty housing provides superior protection, making them ideal for tough applications. Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
All models feature Min/Max/Avg, Hold, °F/°C or Kelvin (K) selectable units, relative time stamp on Min/Max/Avg, electronic offset function, temperature differential, backlit display, and auto power-off. Accept a wide variety of type K thermocouple probes with miniconnector.
Ordering information: These meters also feature an infrared input that accepts an optional IR wand for noncontact measurements of objects that are difficult or dangerous to reach; order wand 20250-24 separately. The IR temperature reading can be used as a unique input when calculating temperature differential. The IR wand has a distance-to sight ratio of 8:1 and temperature range of –35 to 550 °C. For more information; contact your Avantor representative.
Delivery: Four Type K bead wire temperature probes, hard carrying case, USB cable, software, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.