ContactUsLinkComponent
MICROFLEX® 94-242 Nitrile Static Dissipative Gloves
 :  Ansell
undefined
MICROFLEX® 94-242 Nitrile Static Dissipative Gloves
 :  Ansell
  • Catalog No:
  • 112-6423
  • 112-6422
  • 112-6421
  • 112-6420
  • 112-6424
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Textured fingers
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff length:
  • Standard
  • Standard
  • Standard
  • Standard
  • Standard
  • Cuff thickness:
  • 0,08 mm (3,1 mil)
  • 0,08 mm (3,1 mil)
  • 0,08 mm (3,1 mil)
  • 0,08 mm (3,1 mil)
  • 0,08 mm (3,1 mil)
  • Finger thickness:
  • 0,16 mm (6,2 mil)
  • 0,16 mm (6,2 mil)
  • 0,16 mm (6,2 mil)
  • 0,16 mm (6,2 mil)
  • 0,16 mm (6,2 mil)
  • Palm thickness:
  • 0,12 mm (4,7 mil)
  • 0,12 mm (4,7 mil)
  • 0,12 mm (4,7 mil)
  • 0,12 mm (4,7 mil)
  • 0,12 mm (4,7 mil)
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
Frequently Bought Together