Orders LinkContactUsLinkComponent
MICROFLEX® 31-103 Compostable Disposable Gloves
MICROFLEX® 31-103 Compostable Disposable Gloves
Catalog #
MICROFLEX® 31-103 Compostable Disposable Gloves
Catalog #
Supplier:  
MICROFLEX® 31-103 Compostable Disposable Gloves
MICROFLEX® 31-103 Compostable Disposable Gloves
Product Details & Documents
MICROFLEX® 31-103 gloves achieve 90%+ biodegradation within 180 days in industrial composting facilities.
  • Bio-based PLA (polyactic acid) blend formulation reduces environmental impact
  • EN 13432 certification for certified compostable gloves
  • Made in Portugal (EU); supporting local manufacturing
Offering certified protection, they are the only PLA-blend product on the market with EN ISO 374-1 Type C and EN ISO 374-5 virus certifications. These eco-friendly gloves are packed in 200-count compostable polybags for decreased packaging waste. Their manufacturing process uses 100% green energy from solar power and hydroelectricity.
To reduce allergy risks, these gloves do not contain silicones or accelerators and are allergen-free.
Certifications: CE 0493, EN 13432, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, FDA21 CFR 177-2600-US Food Contact Approved, Food Contact, ISO 9001, OK compost INDUSTRIAL (TUV Austria), UKCA 0321.