Orders LinkContactUsLinkComponent
Cleanroom Tunics
Cleanroom Tunics
Supplier:   ALSICO HIGHTECH
Cleanroom Tunics
Supplier:  ALSICO HIGHTECH
Certificates
Cleanroom Tunics
7 Options Available Below
Product Details & Documents
These shirts are made of Movix (96% polyester, 4% carbon). Suitable for C and D classes or ISO 7 to 9 cleanrooms. For higher cleanroom classes worn as an undergarment piece under a coverall. Can also be used for clean areas.
  • V-neck
  • Long inserted sleeves
  • Seamed at waist
Usable in ISO 7 to 9.
Certifications: IEST-RP-CC003.5
Specifications for Product Family
Specifications
  • Catalog No:
  • ALSO1033607-XS
  • ALSO1033607-S
  • ALSO1033607-M
  • ALSO1033607-L
  • ALSO1033607-XL
  • ALSO1033607-XXL
  • ALSO1033607-3XL
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Risk area:
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Movix anti-static (96% polyester, 4% carbon)
  • Weight:
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
  • 152 g/m²
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • Dark blue
    XS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-XS
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
  • Dark blue
    S
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-S
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
  • Dark blue
    M
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-M
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
  • Dark blue
    L
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-L
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
  • Dark blue
    XL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-XL
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
  • Dark blue
    XXL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-XXL
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
  • Dark blue
    3XL
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    ALSO1033607-3XL
    Garment application
    Cleanroom
    Risk area
    Anti-static
    Gender
    Unisex
    Fabric
    Movix anti-static (96% polyester, 4% carbon)
    Weight
    152 g/m²
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.