Orders LinkContactUsLinkComponent
Pharma Scoops
Pharma Scoops
Supplier:   BOCHEM
Pharma Scoops
Supplier:  BOCHEM
Certificates
Pharma Scoops
4 Options Available Below
Product Details & Documents
These scoops are made from V4A steel.
  • Very good chemical resistance
  • Excellent thermal resistance, autoclavable
  • Highly polished
  • With seamless fixed handle
Ideal for sample taking in the pharmaceutical industry.
Product Family Options
Product Information
  • Capacity
    Length
    Width
    Availability
    Price
  • BOCH8860
    Pharma scoop
    Pharma scoop
    100 ml
    100 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
  • BOCH8861
    Pharma scoop
    Pharma scoop
    450 ml
    150 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 180,00 €
    180,00 €
    1 * 1 ST
       180,00 €
  • BOCH8862
    Pharma scoop
    Pharma scoop
    1000 ml
    235 mm
    110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
  • BOCH8863
    Pharma scoop
    Pharma scoop
    2500 ml
    250 mm
    135 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 234,00 €
    234,00 €
    1 * 1 ST
       234,00 €