Orders LinkContactUsLinkComponent
Bioanalyzer High Sensitivity Protein Analysis
Supplier:   AGILENT
Bioanalyzer High Sensitivity Protein Analysis
Supplier:  AGILENT
Certificates
Bioanalyzer High Sensitivity Protein Analysis
4 Options Available Below
Product Details & Documents
High sensitivity electrophoretic separation of proteins and peptides up to 250 kD.
  • A ready-to-use system featuring standardised assays and pre-packaged reagent kits which are suitable to analyse samples up to 250 kD
  • After sample labelling, fast results are achieved with automated electrophoresis of 10 samples in 30 minutes
  • Highly sensitive with an accuracy down to silver stain levels, approximately 1 pg/µl of labelled protein on chip
  • Fully operational across a large linear dynamic range with four orders of magnitude
  • Highly precise impurity detection, down to 0,05% and lower
Perform high sensitivity protein electrophoresis with the 2100 Bioanalyzer system. The high sensitivity protein 250 assay provides a broad dynamic range with sensitivity down to silver stain comparable levels. Efficiency is enhanced by eliminating handling of SDS-PAGE slab gels, staining or, imaging steps.
The system provides a fast, automated, objective and flexible solution for protein and peptide characterisation, quality control, and impurity detection in the low picogram range. Compatible sample types include protein lysates, purified protein and peptides, antibodies, and protein stability tests.
 : For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • Details
    Availability
    Price
  • AGLS5067-1575
    Agilent high sensitivity protein 250 kit
    Agilent high sensitivity protein 250 kit
    For sizing and highly sensitive quantitation of protein samples from 10 - 250 kDa, the kit includes 10 microfluidic chips, reagents and consumables, sufficient for 100 samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 163,65 €
    1 163,65 €
    1 * 1 ST
       1 163,65 €
  • AGLS5067-1577
    Agilent high sensitivity protein 250 labeling kit
    Agilent high sensitivity protein 250 labeling kit
    Reagents for labeling reaction only, sufficient for 100 samples
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 234,00 €
    234,00 €
    1 * 1 ST
       234,00 €
  • AGLS5067-1576
    Agilent high sensitivity protein 250 reagents
    Agilent high sensitivity protein 250 reagents
    Reagents only, no chips included, sufficient for 10 chips
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 342,00 €
    342,00 €
    1 * 1 ST
       342,00 €
  • AGLS5067-1578
    Agilent high sensitivity protein 250 ladder
    Agilent high sensitivity protein 250 ladder
    Ladder only
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 234,00 €
    234,00 €
    1 * 1 ST
       234,00 €