Orders LinkContactUsLinkComponent
Capillary Arrays
Capillary Arrays
Supplier:   AGILENT
Capillary Arrays
Supplier:  AGILENT
Certificates
Capillary Arrays
6 Options Available Below
Product Details & Documents
Capillary arrays for fragment analyser and ZAG systems.
  • Array cartridges are robust and have a long lifespan, keeping your instrument on-task from one project to the next
  • Short arrays provide robust separation for most samples
  • Speed up separation times and increase overall QC throughput with ultra-short arrays
  • Enhance separation resolution and focus nucleic acid analysis with long arrays
The fragment analyser systems offer a selection of capillary arrays to optimize your QC workflow by separating nucleic acids via parallel capillary electrophoresis. Arrays vary in capillary length, providing distinct benefits to a wide range of workflows.
These arrays address separation speed and separation resolution. 'Short' capillary arrays are the most versatile as they are compatible with all consumable kits. Run times can be decreased with 'Ultra-Short' arrays, and 'Long' arrays enhance separation resolution. Some kits have been designed for a specific array for optimal performance.
Caution: For research use only. Not for use in diagnostic procedures.
Documents
Product Family Options
Product Information
  • Details
    Availability
    Price
  • AGLSA2300-12502247
    FA 12-capillary array ultra-short, 22 cm
    FA 12-capillary array ultra-short, 22 cm
    For use with the fragment analyser 5200 system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 077,00 €
    1 077,00 €
    1 * 1 ST
       1 077,00 €
  • AGLSA2300-12503355
    FA 12-capillary array short, 33 cm
    FA 12-capillary array short, 33 cm
    For use with the fragment analyser 5200 system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 077,00 €
    1 077,00 €
    1 * 1 ST
       1 077,00 €
  • AGLSA2300-12505580
    FA 12-capillary array long, 55 cm
    FA 12-capillary array long, 55 cm
    For use with the fragment analyser 5200 system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 077,00 €
    1 077,00 €
    1 * 1 ST
       1 077,00 €
  • AGLSA2300-48503355
    FA 48-capillary array short, 33 cm
    FA 48-capillary array short, 33 cm
    For use with the fragment analyser 5300 system
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 153,00 €
    2 153,00 €
    1 * 1 ST
       2 153,00 €
  • 732-4440
    FA/ZAG 96-capillary array short, 33 cm
    FA/ZAG 96-capillary array short, 33 cm
    For use with the 5300 or 5400 fragment analyser and ZAG DNA analyser systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 946,00 €
    2 946,00 €
    1 * 1 ST
       2 946,00 €
  • 732-4441
    FA/ZAG 96-capillary array long, 55 cm
    FA/ZAG 96-capillary array long, 55 cm
    For use with the 5300 or 5400 fragment analyser and ZAG DNA analyser systems
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 946,00 €
    2 946,00 €
    1 * 1 ST
       2 946,00 €