Order Entry
Belgium
ContactUsLinkComponent
Luciferase cell lysis buffer (2X), Pierce™
Luciferase cell lysis buffer (2X), Pierce™
Catalog # PIER16189
CAS Number:  
undefined
Luciferase cell lysis buffer (2X), Pierce™
Catalog # PIER16189
CAS Number:  

Specifications

  • Pk:
    250 ml
  • Description:
    Luciferase cell lysis buffer, 2X
  • Storage conditions:
    Store at room temperature

Specifications

About this item

The Thermo Scientific Pierce luciferase cell lysis buffer effectively lyses adherent cells or pelleted suspension cells for analysis of promoter expression in luciferase assays.

  • Compatible with all Pierce Luciferase reporter assays
  • Enables rapid and efficient lysis
  • Tested compatibility with multiple mammalian cell lines
  • Compatible with protein assays (e.g., coomassie plus and BCA)

Pierce Luciferase Cell Lysis Buffer has been tested and validated to produce luciferase assay-compatible lysates from a variety of cell types. Essentially complete lysis of adherent cells has been observed with nearly all of the cell types tested to date, including COS-7, NIH3T3, Hepa 1-6, 293, CHO, MDA, MB 231, Jurkat, and FM2 cells.

Caution: For research use only. Not for use in diagnostic procedures.