ContactUsLinkComponent
VWR®, Polypropylene Microplates, 96-Well
VWR®, Polypropylene Microplates, 96-Well
Catalog # 548-3375
Supplier:  VWR Collection
undefined
VWR®, Polypropylene Microplates, 96-Well
Catalog # 548-3375
Supplier:  VWR Collection
Specifications
  • Colour:
    Natural
  • Description:
    Polypropylene deep well microplate, Ø×H: 7×44 mm, well depth: 44 mm, square shaped
  • Pk:
    5
  • Well volume:
    2000 µl
Specifications
About this item
VWR® 96-Well plates and covers are designed for chromatography and high throughput screening (HTS) applications.
  • SBS and ANSI standard footprint
  • Low binding virgin PP for homogenous assays and storage
  • –80 to +121 °C tolerant
VWR® 96-Well Microplates are manufactured specifically for chromatography. Multiple volumes and interior finishes allow for enhanced sample recovery. Low binding virgin polypropylene plates are perfect for high throughput screening (HTS), and sensitive samples.