Supplier: G-Biosciences
Certificates
About this item
Specifications
- Catalog No:
- GENOITM3571-50U
- GENOITM3571-100U
- Antigen symbol:
- Immunotag™ Kir4.1
- Immunotag™ Kir4.1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic Peptide of Kir4.1
- Synthetic Peptide of Kir4.1
- Form:
- Liquid
- Liquid
- Purification:
- The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using specific immunogen
- The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using specific immunogen
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide.
- PBS containing 50% glycerol, 0.5% BSA and 0.02% sodium azide.
- Storage temperature:
- -20 °C
- -20 °C
- Accession no.:
- P78508 Q9JM63 P49655
- P78508 Q9JM63 P49655
Specifications
Frequently Bought Together