Orders LinkContactUsLinkComponent
Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable, 6' cable
Catalog # MFLX07526-99
Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Catalog # MFLX07526-99
Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Masterflex® Universal Pressure Sensor Adapter Cable, Avantor®
Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable, 6' cable
Price
153,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Adapts a variety of sensors to the MasterSense™ Sensor AUX input.
  • Designed for use with 0-20/4-20 mA or 0-5,0-10 V sensors
  • Includes 24VDC power out pins for loop powered circuits up to 50 mA
Connection cable allows for integration of variety of sensors into Masterflex® MasterSense™ pump systems. Reference MasterSense™ instruction manual for wiring diagrams and detail.
Specifications
  • Description:
    Masterflex® universal pressure sensor adaptor cable, bare wire- MasterSense™ pump systems, 6' cable
  • Included accessories:
    6' cable
  • Cat. No.:
    MFLX07526-99
  • Max. pressure:
    100 psi