Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
92498.320
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Assay :min. 99.9%
- Water :max. 0.02%
- Evaporation Residue:max. 0.0001%
- Acidity [meq/g]:max. 0.0003
- Alkalinity [meq/g]:max. 0.0002
- Gradient use test (230 nm):max. 0.002 AU
- Gradient use test (254 nm):max. 0.002 AU
- Baseline drift (230 nm):max. 0.02 AU
- Fluorescence (as quinine; 254 nm):max. 1 ppb
- Fluorescence (as quinine; 365 nm):max. 1 ppb
- UV transmission (205 nm):min. 10%
- UV transmission (210 nm):min. 50%
- UV transmission (225 nm):min. 80%
- UV transmission (240 nm):min. 95%
- UV transmission (265-400 nm):min. 99%
- Ag (Silver):max. 10 ppb
- Al (Aluminium):max. 25 ppb
- Ca (Calcium):max. 25 ppb
- Cu (Copper):max. 10 ppb
- Fe (Iron):max. 10 ppb
- K (Potassium):max. 10 ppb
- Mg (Magnesium):max. 10 ppb
- Mn (Manganese):max. 10 ppb
- Na (Sodium):max. 50 ppb
- Ni (Nickel):max. 10 ppb
- Pb (Lead):max. 10 ppb
- Zn (Zinc):max. 10 ppb
- Suitability for PFAS analysis:passes test
- Suitability for LC-MS:passes test
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:13,05984
:64.6 °C (1013 hPa)
:–98 °C
:0.7918 g/cm³ (20 °C)
:Ambient
:200-659-6
:1230
:3 (6.1),II
:Packaged under nitrogen.
:11 °C (closed cup)
:01-2119433307-44
:GHS02
:GHS06
:GHS08