Orders LinkContactUsLinkComponent
VWR®, Cryo Boxes
VWR®, Cryo Boxes
VWR®, Cryo Boxes
VWR®, Cryo Boxes
VWR®, Cryo Boxes
Cryo boxes are suitable to store up to –196 °C.
  • Suitable for storage temperatures down to –196°C
  • Available in a variety of color options
  • Grid option for coding on lid
  • Clear lid material
Made with PC material
Boxes are stackable. Designed for 25, 81, or 100 places to store both internal and external threaded vials (1 - 5 ml).
Base made with color options red, blue, green, white, and yellow. Lid has grid option for coding and made with clear material.
Ordering information: Cryo tube picker is included.
Product Family Options
  • Compartments
    Colour
    For tubes
    W×D×H ext.
  • 25
    Assorted
    1 - 2 ml
    75,6×75,6×52,2 mm
    1 * 5 ST
    1 * 5 ST - 43,30 €
    43,30 €
    1 * 5 ST
       43,30 €
  • 25
    Blue
    1 - 2 ml
    75,6×75,6×52,2 mm
    1 * 5 ST
    1 * 5 ST - 43,30 €
    43,30 €
    1 * 5 ST
       43,30 €
  • 25
    Green
    1 - 2 ml
    75,6×75,6×52,2 mm
    1 * 5 ST
    1 * 5 ST - 43,30 €
    43,30 €
    1 * 5 ST
       43,30 €
  • 25
    Red
    1 - 2 ml
    75,6×75,6×52,2 mm
    1 * 5 ST
    1 * 5 ST - 43,30 €
    43,30 €
    1 * 5 ST
       43,30 €
  • 25
    White
    1 - 2 ml
    75,6×75,6×52,2 mm
    1 * 5 ST
    1 * 5 ST - 43,30 €
    43,30 €
    1 * 5 ST
       43,30 €
  • 25
    Yellow
    1 - 2 ml
    75,6×75,6×52,2 mm
    1 * 5 ST
    1 * 5 ST - 43,30 €
    43,30 €
    1 * 5 ST
       43,30 €
  • 81
    Assorted
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,70 €
    69,70 €
    1 * 5 ST
       69,70 €
  • 81
    Blue
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,70 €
    69,70 €
    1 * 5 ST
       69,70 €
  • 81
    Green
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,70 €
    69,70 €
    1 * 5 ST
       69,70 €
  • 81
    Red
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,70 €
    69,70 €
    1 * 5 ST
       69,70 €
  • 81
    White
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,70 €
    69,70 €
    1 * 5 ST
       69,70 €
  • 81
    Yellow
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,70 €
    69,70 €
    1 * 5 ST
       69,70 €
  • 81
    Assorted
    3 ml
    132×132×81,4 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Blue
    3 ml
    132×132×81,4 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Green
    3 ml
    132×132×81,4 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Red
    3 ml
    132×132×81,4 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    White
    3 ml
    132×132×81,4 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Yellow
    3 ml
    132×132×81,4 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Assorted
    4 - 5 ml
    132×132×94,3 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Blue
    4 - 5 ml
    132×132×94,3 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Green
    4 - 5 ml
    132×132×94,3 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Red
    4 - 5 ml
    132×132×94,3 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    White
    4 - 5 ml
    132×132×94,3 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 81
    Yellow
    4 - 5 ml
    132×132×94,3 mm
    1 * 5 ST
    1 * 5 ST - 68,00 €
    68,00 €
    1 * 5 ST
       68,00 €
  • 100
    Assorted
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,10 €
    69,10 €
    1 * 5 ST
       69,10 €
  • 100
    Blue
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,10 €
    69,10 €
    1 * 5 ST
       69,10 €
  • 100
    Green
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,10 €
    69,10 €
    1 * 5 ST
       69,10 €
  • 100
    Red
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,10 €
    69,10 €
    1 * 5 ST
       69,10 €
  • 100
    White
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,10 €
    69,10 €
    1 * 5 ST
       69,10 €
  • 100
    Yellow
    1 - 2 ml
    132×132×53 mm
    1 * 5 ST
    1 * 5 ST - 69,10 €
    69,10 €
    1 * 5 ST
       69,10 €
Cryoboxes for 2 ml 2D barcoded cryovials
  • Compartments
    Colour
    For tubes
    W×D×H ext.
  • 81
    Blue
    2 ml
    1 * 5 ST
    1 * 5 ST - 52,30 €
    52,30 €
    1 * 5 ST
       52,30 €
  • 81
    Green
    2 ml
    1 * 5 ST
    1 * 5 ST - 52,30 €
    52,30 €
    1 * 5 ST
       52,30 €
  • 81
    Red
    2 ml
    1 * 5 ST
    1 * 5 ST - 52,30 €
    52,30 €
    1 * 5 ST
       52,30 €
  • 81
    Yellow
    2 ml
    1 * 5 ST
    1 * 5 ST - 52,30 €
    52,30 €
    1 * 5 ST
       52,30 €
Accessories
  •  
    Description
  • 479-2294
    CryoHolder, model T
    1 * 1 ST