ContactUsLinkComponent
Performance Professional Refrigerators with Dynamic cooling and Plastic Inner Liner, FRFvg
 :  Liebherr
undefined
Performance Professional Refrigerators with Dynamic cooling and Plastic Inner Liner, FRFvg
 :  Liebherr
  • Catalog No:
  • LIEBFRFVG6501
  • LIEBFRFVG5501
  • LIEBFRFVG4001
  • Model:
  • FRFvg 6501
  • FRFvg 5501
  • FRFvg 4001
  • Display:
  • Monochrome display, touch buttons, external digital
  • Monochrome display, touch buttons, external digital
  • Monochrome display, touch buttons, external digital
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Alarm:
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Shelves/drawers:
  • 5 adjustable, wire shelves plastic-coated
  • 4 adjustable, wire shelves plastic-coated
  • 5 adjustable, wire shelves plastic-coated
  • Door type:
  • Steel, self closing
  • Steel, self closing
  • Steel, self closing
  • Door lock:
  • Mechanical
  • Mechanical
  • Mechanical
  • Insulation:
  • 45/57 mm
  • 45/57 mm
  • 45/57 mm
  • Refrigerant:
  • R 600a
  • R 600a
  • R 600a
  • Sound level:
  • 49 dB(A)
  • 49 dB(A)
  • 48 dB(A)
  • Energy class:
  • C
  • C
  • C
  • Electrical:
  • 220 - 240 V~, 2,0 A, 50 Hz
  • 220 - 240 V~, 2,0 A, 50 Hz
  • 220 - 240 V~, 2,0 A, 50 Hz
  • Weight:
  • 93 (gross)/84 (net) kg
  • 87 (gross)/78 (net) kg
  • 70 (gross)/65 (net) kg
Frequently Bought Together