ContactUsLinkComponent
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
 :  Liebherr
undefined
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
 :  Liebherr
  • Catalog No:
  • LIEBHMFVH4011
  • LIEBHMFVH4001H63
  • LIEBHMFVH5501
  • LIEBHMFVH4011H63
  • LIEBHMFVH5511
  • LIEBHMFVH5501H63
  • LIEBHMFVH5511H63
  • LIEBHMFVH4001
  • Model:
  • HMFvh 4011
  • HMFvh 4001 H63
  • HMFvh 5501
  • HMFvh 4011 H63
  • HMFvh 5511
  • HMFvh 5501 H63
  • HMFvh 5511 H63
  • HMFvh 4001
  • Display:
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • 2,4" touch and swipe colour display
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Alarm:
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Visual and audible
  • Access ports:
  • 1ר 10 mm
  • 1ר 10 mm
  • 1ר 10 mm
  • 1ר 10 mm
  • Shelves/drawers:
  • 6 (5 adjustable), wire shelves plastic-coated
  • 1 adjustable, wire shelf plastic-coated/8 drawers
  • 6 (5 adjustable), wire shelves plastic-coated
  • 1 adjustable, wire shelf plastic-coated/8 drawers
  • 6 (5 adjustable), wire shelves plastic-coated
  • 1 adjustable, wire shelf plastic-coated/7 drawers
  • 1 adjustable, wire shelf plastic-coated/7 drawers
  • 6 (5 adjustable), wire shelves plastic-coated
  • Door type:
  • glass
  • steel
  • steel
  • glass
  • glass
  • steel
  • glass
  • steel
  • Door lock:
  • electronic, with remote control
  • electronic, with remote control
  • electronic, with remote control
  • electronic, with remote control
  • electronic, with remote control
  • electronic, with remote control
  • electronic, with remote control
  • electronic, with remote control
  • Refrigerant:
  • R 600a
  • R 600a
  • R 600a
  • R 600a
  • R 600a
  • R 600a
  • R 600a
  • R 600a
  • Lamp:
  • Vertical LED light strip recessed in left wall with separate switch
  • Vertical LED light strip recessed in left wall with separate switch
  • LED lighting column, left
  • Vertical LED light strip recessed in left wall with separate switch
  • Vertical LED light strip recessed in left wall with separate switch
  • Vertical LED light strip recessed in left wall with separate switch
  • Vertical LED light strip recessed in left wall with separate switch
  • LED lighting column, left
  • Sound level:
  • 49 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • 49 dB(A)
  • Electrical:
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • 220 - 240V~, 2,0 A
  • Weight:
  • 89 (gross)/83 (net) kg
  • 69 (gross)/64,5 (net) kg
  • 91 (gross)/82 (net) kg
  • 82 (gross)/69 (net) kg
  • 111 (gross)/102 (net) kg
  • 106 (gross)/100 (net) kg
  • 101 (gross)/97 (net) kg
  • 73 (gross)/64,5 (net) kg
Frequently Bought Together