ContactUsLinkComponent
Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Certificates
undefined
Masterflex® Nylon Clamps for Sanitary Fittings, Avantor®
Specifications
  • Sanitary clamp size (port 1):
  • 2"
  • 1/2"
  • 1-1/2"
  • Sterilisation:
  • Autoclave, Gamma
  • Autoclave, Gamma
  • Autoclave, Gamma