Orders LinkContactUsLinkComponent
Rigid Conductive Boxes
Rigid Conductive Boxes
Supplier:   DESCO
Rigid Conductive Boxes
Supplier:  DESCO
Certificates
Rigid Conductive Boxes
3 Options Available Below
Product Details & Documents
Desco Europe Rigid Conductive Boxes are marked with the ESD protective symbol. Boxes can be supplied with or without high density foam for insertion of component leads or low density foam which acts as a cushioning material.
  • Provide good ESD and mechanical protection
  • Made of conductive polypropylene; Rs: 1×10² to <1×10⁵ Ω per IEC 61340-2-3
  • Strong box moulded in one-piece
  • Supplied with/without foam lining
  • Top foam: 6 mm thick low density foam (FX)
  • Bottom foam: 6 mm thick high density foam (HD)
Certifications: Meets EN 61340-5-1 and Packaging standard IEC 61340-5-3.
Product Family Options
With foam lining
Product Information
  • Availability
    Price
  • 129-2620
    Rigid conductive box with foam in lid and base, 75×51×14 mm
    Rigid conductive box with foam in lid and base, 75×51×14 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,10 €
    4,10 €
    1 * 1 ST
       4,10 €
  • 129-2619
    Rigid conductive box with foam in lid and base, 90×64×16 mm
    Rigid conductive box with foam in lid and base, 90×64×16 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5,00 €
    5,00 €
    1 * 1 ST
       5,00 €
Without foam lining
Product Information
  • Availability
    Price
  • 129-2621
    Rigid conductive box, 230×128×30 mm
    Rigid conductive box, 230×128×30 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7,10 €
    7,10 €
    1 * 1 ST
       7,10 €