Orders LinkContactUsLinkComponent
Conductive Tote Boxes
Conductive Tote Boxes
Supplier:   DESCO
Conductive Tote Boxes
Supplier:  DESCO
Certificates
Conductive Tote Boxes
2 Options Available Below
Product Details & Documents
Conductive tote boxes are made from injection moulded conductive polypropylene.
  • Flat base, stackable
  • Rv: 1×10³ to <1×10⁶ Ω
  • Custom-made dividers are available for PCB containers
  • Machined from approximately 2 mm thick conductive plastic or Cortronic® B-flute static dissipative cardboard
  • Many shapes and sizes possible
  • Process is suited for small and medium quantities
Product Family Options
Product Information
  • Availability
    Price
  • DESC239305
    Conductive tote box, 400×300×120 mm
    Conductive tote box, 400×300×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,40 €
    27,40 €
    1 * 1 ST
       27,40 €
  • DESC239345
    Conductive tote box, 600×400×236 mm
    Conductive tote box, 600×400×236 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,90 €
    69,90 €
    1 * 1 ST
       69,90 €