Orders LinkContactUsLinkComponent
Static dissipative facemasks
Static dissipative facemasks
Supplier:   DESCO
Static dissipative facemasks
Supplier:  DESCO
Certificates
Static dissipative facemasks
6 Options Available Below
Product Details & Documents
These facemasks constructed of a lightweight dissipative material which is made from texturised polyester and a minimum of 9% carbon nylon monofiliment.
  • Can be washed multiple times in a mild detergent
  • Elastic ear loops
  • Adjustable nose wire helps ensure mask fits securely
  • Acceptable for use in ESD protected areas
  • Provides continuous and consistent charge dissipation
Elastic bands range from small-medium (69,85 mm) to large-extra large (82,55 mm). The mask area is the same size for all items.
Product Family Options
Product Information
  • Size
    Colour
    Availability
    Price
  • S/M
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,80 €
    22,80 €
    1 * 1 ST
       22,80 €
  • S/M
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,40 €
    23,40 €
    1 * 1 ST
       23,40 €
  • S/M
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,40 €
    23,40 €
    1 * 1 ST
       23,40 €
  • L/XL
    Blue
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,40 €
    23,40 €
    1 * 1 ST
       23,40 €
  • L/XL
    White
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,40 €
    23,40 €
    1 * 1 ST
       23,40 €
  • L/XL
    Grey
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,40 €
    23,40 €
    1 * 1 ST
       23,40 €