ContactUsLinkComponent
Disposable conductive booties
Disposable conductive booties
Catalog # 114-9321
Supplier:  DESCO
Disposable conductive booties
Catalog # 114-9321
Supplier:  DESCO
Specifications
  • Anti-static:
    +
  • Colour:
    Blue
  • Description:
    Disposable conductive booties
  • Pk:
    150 PAIR
Specifications
About this item
Disposable conductive surgical style Booties are 'one size fits all' with elastic opening.
  • Lightweight
  • Cleanroom compatible
  • Maintains grounding with full mobility
  • Will not scratch or mark flooring
Disposable ESD Booties can be used for person/footwear/flooring system per EN 61340-5-1.
Packaging: Sold 150 pairs per box.