Silicone lubricant and sealant, Molykote™ 111
636057A
- Description:Molykote™111
- Environmentally Preferable:
- Pk:5 kg
Molykote 111 is a valve lubricant and sealant with good resistance to most chemicals and a wide service temperature range.
- Wide service temperature range (40 to 200 °C)
- Low vapour pressure
- Low volatility
- Excellent water resistance