Microplate washer, HydroFlex™ Plus
Product Details & Documents
The HydroFlex Plus microplate washer is a truly flexible and upgradeable platform for automated microplate strip washing n 96-well plate formats. It is an ideal solution for a wide range of cell- and ELISA-based applications.
- Control of wash head positions and speed settings for gentle cell washing
- Simultaneous aspiration and dispensing for ELISA washing
- Up to two aspiration points per well for minimal residual volumes
- Ready for robotic integration
- Automated buffer switching of four liquids and plate shaking capability
- Predefined plate library and protocols for an easy start
- Designed to meet IVDR (EU) 2017/746 (only for ELISA wash)
The HydroFlex Plus offers both cell and ELISA washing, without the need for dedicated wash heads. It provides consistent, high- quality washing with very low residual volumes of less than 2 μl per well, and features multiple aspiration points for flat-bottom well geometries.
The overflow function offers excellent single-step washing effciency, avoiding the need for tedious repositioning of the wash head for sequential aspiration and dispense steps.
: Accessories not included (must be ordered seperate). Additional configuration available on request.
: Includes 1× Washer, 1× 5 L set bottle waste, 2× 2,5 L wash bottles, 1× power cable, 1× USB stick with manuals in 11 languages.
Documents
Specifications
- Description:Microplate washer for 96 well plates
- Seq:01
- Dispensing volume:50 - 400 µl in 50 µl increments
- Wash/waste bottle capacity:2×2,5L wash bottles, 1×5 L waste bottle
- Wash heads:1×8
- Shaking:Linear in three different intensities
- Weight:6,6 kg
- Cat. No.:TCAT91100001
- Dispensing precision:≤2,5 % CV uniformity
- Model:HydroFlex Plus
- W×D×H:275×366×180,5 mm
- Plate types:96-well plates
- Priming and rinsing volume:15 - 820 ml
- Washing volume:50 - 3000 µl in 50 µl increments
- Residual aspiration volume:<2 µl (flat bottom plates)
<5 µl (V-shaped bottom plates)
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



