Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microplate reader, Sunrise®

Microplate reader, Sunrise®

Sunrise, Singlemode microplate reader for measuring of absorbance with data analysis software Magellan V7.5 STD 2PC
Catalog # TCAT95000001
Supplier:   Tecan
Microplate reader, Sunrise®
Catalog # TCAT95000001
Supplier:  Tecan
Microplate reader, Sunrise®
Microplate reader, Sunrise®

Sunrise, Singlemode microplate reader for measuring of absorbance with data analysis software Magellan V7.5 STD 2PC

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

This Sunrise absorbance microplate reader offers the functionality needed for numerous photometric applications.

  • Ideal for colometric ELISA, kinetic assays and agglutination readings
  • Temperature control option for enzyme kinetics
  • Advanced 12-channel optics + reference channel for fast and optimised results
  • Microplate shaking
  • Ready for robotic integration

The Sunrise absorbance reader, complemented by Tecan’s Magellan™ universal reader control and data analysis software add-on, is an ideal solution for a range of applications, such as ELISAs, enzyme kinetics, cell viability studies, veterinary assays and many more.

It supports a broad wavelength range – from 340 to 750 nm – and can measure an entire 96-well plate in less than si× seconds, ensuring it is ready to meet the needs of routine research labs. The three-year warranty demonstrate Tecan’s commitment to quality.

Ordering information: Accessories not included (must be ordered seperately). Additional configuration available on request.

Delivery information: Includes 1× reader, 1× data analysis software 1× power cable, 1× USB stick with manuals in 11 languages.

Specifications
  • Model:
    Sunrise
  • Description:
    Singlemode microplate reader for measuring of absorbance with data analysis software Magellan V7.5 STD 2PC
  • Seq:
    01
  • Light source:
    Halogen lamp 20 W
  • Shaking:
    Linear shaking in four different modes
  • Weight:
    ma×. 8,6 kg (incl all options)
  • Interfaces:
    Serial RS232C (remote control)
  • Cat. No.:
    TCAT95000001
  • Filters:
    Four-filter optics
  • Temperature operating range:
    15...35 °C
  • Wavelength selection:
    12-channel optics + reference channel
  • Bandwidth:
    10 ±2 nm
  • Wavelength range:
    340 - 750 nm
  • W×D×H:
    285×340×145 mm
  • Plate types:
    96-well plates
  • Temperature control:
    Room temperature up to 42 °C increments of 0,1 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.