ContactUsLinkComponent
Union Fittings, Hose Barb, Cross Connector, Dynalon®
undefined
Union Fittings, Hose Barb, Cross Connector, Dynalon®
  • Orifice dimensions:
  • 8 mm (0.3125")
  • 9 mm (0.359375")
  • 6 mm (0.234375")
  • 2 mm (0.078125")
  • 4 mm (0.15625")
  • Sterilisation:
  • Autoclave only
  • Autoclave only
  • Autoclave only
  • Autoclave only
  • Autoclave only
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together