Quanti-CultiControl freeze-dried microorganism strains derived from ATCC® cultures
Supplier: LIOFILCHEM
Certificates
About this item
Quanti-CultiControl are lyophilized, quantitative microorganism preparations to be used in industrial laboratories for Quality Control purposes. Processed as directed, these preparations provide a challenge of < 100 CFU per 0.1 mL, a concentration suitable for several pharmaceutical applications, including growth promotion testing of culture media to be employed in sterility testing
- Derived from the American Type Culture Collection (ATCC®)
- Suitable for the Growth Promotion Test recommended by the international Pharmacopoeiae
- No need of intermediate dilution steps, since each rehydrated suspension generates less than 100 UFC for 0.1 ml inoculum
- 24 different Recommended Growth Methods are available (see the technical document in the documentation sheet)
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Liofilchem® s.r.l. is licensed to use these trademarks and to sell products derived from ATCC® cultures
Certifications: Suitable for the Growth Promotion Test recommended by the International Pharmacopoeiae.
Packaging: 1 vial containing 1 pellet + 1 rehydration fluid vial. Each suspension (1 ml) allows 10 inocula.
Frequently Bought Together