Order Entry
Belgium
ContactUsLinkComponent
Ethanol absolute ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Flammable
Irritant
Ethanol absolute ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
Ethanol absolute ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    ≥ 99.9 %
  • Acidity:
    ≤ 0.0001 meq/g
  • Alkalinity:
    ≤ 0.0001 meq/g
  • Colouration:
    ≤ 10 APHA
  • Evaporation residue:
    ≤ 2 ppm
  • Water:
    ≤ 0.05 %
  • Absorbance (207 nm):
    ≤ 0.70
  • Absorbance (220 nm):
    ≤ 0.26
  • Absorbance (235 nm):
    ≤ 0.10
  • Absorbance (240 nm):
    ≤ 0.07
  • Absorbance (245 nm):
    ≤ 0.05
  • Absorbance (from 260 nm):
    ≤ 0.01
  • Transmittance (207 nm):
    ≥ 20 %
  • Transmittance (220 nm):
    ≥ 55 %
  • Transmittance (235 nm):
    ≥ 80 %
  • Transmittance (240 nm):
    ≥ 85 %
  • Transmittance (245 nm):
    ≥ 90 %
  • Transmittance (from 260 nm):
    ≥ 98 %
  • Fluorescence (as quinine) (254 nm):
    ≤ 1.0 ppb
  • Fluorescence (as quinine) (365 nm):
    ≤ 1.0 ppb

Specification Test Results

Related Information

Frequently Bought Together