Order Entry
Belgium
ContactUsLinkComponent
VWR®, Mixed Anions Standard, 7 components in H₂O
VWR®, Mixed Anions Standard, 7 components in H₂O
Catalog # 91295.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Mixed Anions Standard, 7 components in H₂O
Catalog # 91295.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pack type:
    HDPE bottle
  • Pk:
    100 mL
  • Description:
    Mixed Anions Standard (7 components)
  • Seq:
    02
  • Storage temperature:
    Ambient
  • Elements and Concentration:
    100 mg/l each of Chlorides (Cl-), Fluorides (F-), Sulphates (SO42-), Nitrates (NO3-), Nitrites (NO2-), Bromides (Br-), Phosphates (PO43-)

Specifications

About this item

7 components: 100 mg/l each of Chlorides (Cl-), Fluorides (F-), Sulphates (SO42-), Nitrates (NO3-), Nitrites (NO2-), Bromides (Br-), Phosphates (PO43-) in H₂O

Certifications: ISO 17025/17034