ContactUsLinkComponent
Pleated masks with earloops, non sterile, HM4, PUREZERO®
Pleated masks with earloops, non sterile, HM4, PUREZERO®
Catalog # 112-6507
Supplier:  Halyard
undefined
Pleated masks with earloops, non sterile, HM4, PUREZERO®
Catalog # 112-6507
Supplier:  Halyard
Specifications
  • Type:
    Pleated masks with earloops, non-sterile
  • Colour:
    White
  • Pk:
    500
Specifications
About this item
The HM4 Pleated Mask with Earloops is a three-layer mask with nylon earloops that provides a high particle, bacterial, and viral filtration efficiency. The mask construction consists of low-linting materials and a polyester foam nose band.
  • Three-layer mask
  • Nylon earloops
  • Designed for industrial and cleanroom use
  • Recommended for ISO Class 4 and or higher and Grade B/C/D cleanrooms
  • Exclusive SO SOFT biocomponent technology inner lining for comfort
Cleanroom efficiency depends on effective risk management. Ensure masks are up to the task with PUREZERO® HM4 non-sterile face masks, which are specifically designed to meet the stringent requirements of cleanroom environments. This three-layer mask features a unique bicomponent material (polyester and polyethylene). They are manufactured in facilities with raw materials and components traceable to the original supplier.
Packaging: Air pulled, double-bagged. 50 masks per bag, 10 bags per case.