ContactUsLinkComponent
Pleated masks with ties, non sterile, HM4, PUREZERO®
Pleated masks with ties, non sterile, HM4, PUREZERO®
Catalog # 112-6508
Supplier:  Halyard
undefined
Pleated masks with ties, non sterile, HM4, PUREZERO®
Catalog # 112-6508
Supplier:  Halyard
Specifications
  • Type:
    Pleated masks with ties, non sterile
  • Colour:
    White
  • Pk:
    500
Specifications
About this item
The HM4 Pleated Mask with Ties is a three-layer mask with long ties that provides high particle, bacterial, and viral filtration efficiency. This mask is for industrial and cleanroom use. The mask construction consists of low-linting materials with a SO SOFT inner layer, a polyester foam nose band for added comfort, and anti-fog performance.
  • Three-layer mask
  • Long ties for easy donning
  • Designed for industrial and cleanroom use
  • Recommended for ISO Class 4 or higher and Grade B/C/D cleanrooms
  • Exclusive SO SOFT biocomponent technology inner lining for comfort
Cleanroom efficiency depends on effective risk management. Ensure masks are up to the task with PUREZERO® HM4 non sterile face masks, which are specifically designed to meet the stringent requirements of cleanroom environments. This mask features a unique bicomponent material (polyester and polyethylene). They are manufactured in facilities with raw materials and components traceable to the original supplier.
Packaging: Air pulled, double-bagged. 50 masks per bag, 10 bags per case.