ContactUsLinkComponent
Temperature and humidity logger, EBI 20-TH1
Temperature and humidity logger, EBI 20-TH1
Supplier:  EBRO
Certificates
undefined
Temperature and humidity logger, EBI 20-TH1
Supplier:  EBRO
Specifications
  • Catalog No:
  • 620-2837
  • 620-2836
  • Model:
  • EBI 20-TH1
  • EBI 20-TH1
  • Sensor:
  • NTC
  • NTC
  • Protection class:
  • IP 52
  • IP 52
  • Weight:
  • 45 g
  • 45 g
  • W×D×H:
  • 69×48×22 mm
  • 69×48×22 mm
  • Data memory - No. of readings:
  • 40 000
  • 40 000
  • Batteries:
  • 3 V Lithium
  • 3 V Lithium