ContactUsLinkComponent
X5 safety helmet standard reflective sticker kit, SecureFit™
X5 safety helmet standard reflective sticker kit, SecureFit™
  MMMAX5-REFKIT
 :  3M
undefined
X5 safety helmet standard reflective sticker kit, SecureFit™
  MMMAX5-REFKIT
 :  3M
  • Colour:
    Silver
  • Description:
    X5 safety helmet standard reflective sticker kit
  • Pk:
    1
The 3M™ X5-REFKIT reflective sticker kit is designed for use on the 3M SecureFit™ safety helmet series X5000 and X5500.
  • Increase visibility in low-light conditions
  • Pressure sensitive adhesive backed stickers
  • 3M™ Scotchlite reflective material
This kit contains five pieces of pressure sensitive backed 3M™ Scotchlite reflective material to help increase visibility in low light conditions. The kit is designed to fit the 3M SecureFit™ safety helmet series X5000 and X5500.