Advanced polyethylene CentriStar™ cap for centrifuge tubes, Corning®
Supplier: Corning
Certificates
About this item
The CentriStar™ Cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Specifications
- Catalog No:
- 525-0480
- 525-0478
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa