Culture media, diluting and rinsing sterile solutions for sterility testing of pharmaceutical products by membrane filtration.
- Store at 10 to 25 °C away from light
- Shelf life: 2 years
- Packaging, formulation and quality control suitable for media fill trial in pharmaceutical industries
- Some products are available with double or triple irradiated wrapping
Fluid A is a washing solution for rinsing membranes during sterility testing.
Fluid D is a washing solution recommended for dissolving oils or materials containing lecithin or a preservative and can be used for devices labeled as 'sterile pathway'.
Fluid K is a sterile diluent used for rinsing membranes during sterility and is recommended for use with substances containing petrolatum/oil.
Fluid thioglycollate medium and Tryptic soy broth are general purpose liquid enrichment culture media used for sterility control of pharmaceutical products and for cultivation and isolation of fastidious anaerobic and aerobic microorganisms.
TSB is also available as triple-wrapped and gamma-irradiated bottles, particularly suitable for use in restricted areas and for aseptic process simulations (media fill trial) in the pharmaceutical industry.
- Environmentally Preferable: