Ointment jars, with thread
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
About this item
Made from clear or amber soda-lime glass.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- With round thread
Ideal for sampling, storage and transportation of powders and pastes. Amber jars to protect light-sensitive materials.
Ordering information: Screw caps have to be ordered separately.
4 Options Available Below
Product Details & Documents
Made from clear or amber soda-lime glass.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- With round thread
Ideal for sampling, storage and transportation of powders and pastes. Amber jars to protect light-sensitive materials.
Ordering information: Screw caps have to be ordered separately.
Product Family Options
Clear glass
Product Information
Amber glass
Product Information
Screw caps for ointment jars
Product Information
- Catalog #ColourThreadForDescriptionAvailabilityPrice
Accessories
Product Information
- Catalog #AvailabilityPrice
- White polypropylene insert lid for 30 ml jars with thread 46White polypropylene insert lid for 30 ml jars with thread 46RIXI10-23-41Log in for availability1 * 10 ST - 1,80 €