Orders LinkContactUsLinkComponent
Internal thread cryogenic vials
   Sustainable Options Available
Supplier:   Corning
Internal thread cryogenic vials
Supplier:  Corning
   Sustainable Options Available
Certificates
Internal thread cryogenic vials
14 Options Available Below
Product Details & Documents
These vials are manufactured from polypropylene to withstand temperatures down to –196 °C.
  • Black graduations with large marking spot
  • Vials have a silicone washer for a secure seal
  • 2 ml self-standing vials are available in five cap colors.
  • Vials may be color coded with inserts
  • Free foam rack with each case
  • RNase-/DNase-free
  • Sterile
  • Nonpyrogenic
Self-standing vials have a special base design allowing them to be locked into cryogenic rack and tray (Cat. No. 431131) for single-handed manipulation.
 : Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen. Only store vials in the vapor phase above the liquefied gas. Always use appropriate safety equipment when removing vials from cryogenic storage.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-1834
  • 734-4196
  • 734-1835
  • 734-1836
  • 734-1838
  • 734-1837
  • 734-1853
  • 734-1839
  • 479-1187
  • 479-1188
  • 479-1189
  • 479-1190
  • 479-1191
  • 479-1192
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
  • Capacity
    Base
    Cap included
    Cap colour
    Availability
    Price
  • Self standing
    1,20 ml
    Conical bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 360,00 €
    360,00 €
    1 * 500 ST
       360,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1834
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 182,00 €
    182,00 €
    1 * 250 ST
       182,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-4196
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 360,00 €
    360,00 €
    1 * 500 ST
       360,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1835
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 734-1836
    Not self standing
    Not self standing
    2,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 360,00 €
    360,00 €
    1 * 500 ST
       360,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1836
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    4,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 460,00 €
    460,00 €
    1 * 500 ST
       460,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1838
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 734-1837
    Not self standing
    Not self standing
    4,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 444,00 €
    444,00 €
    1 * 500 ST
       444,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1837
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    5,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 460,00 €
    460,00 €
    1 * 500 ST
       460,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1853
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • 734-1839
    Not self standing
    Not self standing
    5,00 ml
    Round bottom
    Yes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 460,00 €
    460,00 €
    1 * 500 ST
       460,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-1839
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Assorted
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 413,00 €
    413,00 €
    1 * 500 ST
       413,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1187
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 378,00 €
    378,00 €
    1 * 500 ST
       378,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1188
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 378,00 €
    378,00 €
    1 * 500 ST
       378,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1189
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 378,00 €
    378,00 €
    1 * 500 ST
       378,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1190
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    Red
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 378,00 €
    378,00 €
    1 * 500 ST
       378,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1191
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
  • Self standing
    2,00 ml
    Round bottom
    Yes
    White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 378,00 €
    378,00 €
    1 * 500 ST
       378,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1192
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
      Low Manufacturing Impact
Cap inserts
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 46,10 €
    46,10 €
    1 * 500 ST
       46,10 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 46,10 €
    46,10 €
    1 * 500 ST
       46,10 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Red
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 46,10 €
    46,10 €
    1 * 500 ST
       46,10 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 46,10 €
    46,10 €
    1 * 500 ST
       46,10 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 46,10 €
    46,10 €
    1 * 500 ST
       46,10 €
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Assorted
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 47,10 €
    47,10 €
    1 * 500 ST
       47,10 €