Orders LinkContactUsLinkComponent
Bar coded external and internal threaded polypropylene cryogenic vials, Corning®
 :   Corning
Bar coded external and internal threaded polypropylene cryogenic vials, Corning®
 :  Corning
Bar coded external and internal threaded polypropylene cryogenic vials, Corning®
Bar coded external and internal threaded polypropylene cryogenic vials, Corning®
These PP cryogenic vials with PP cap protect valuable cells, biological solutions, and aqueous solutions in ultra-low temperature storage.
  • 2 ml capacity, round bottom vials
  • Unique 1D and 2D barcodes, and human readable characters
  • Manufactured from polypropylene to withstand temperatures to as low as –196 °C (–321 °F)
  • Caps designed for a secure seal, and are compatible with automatic decapping equipment
  • Locking base
  • Sterile to SAL 10⁻⁶
  • Certified nonpyrogenic and DNase/RNase-free
The accessory cyrogenic vial boxes are designed for use with 1D and 2D barcoded vials. The 81-Place box accommodates internal or external threaded vials. The 100-Place box accommodates internal threaded vials. Boxes are autoclavable.
Product Family Options
  • Description
    Capacity
    Base
    Cap included
    Cap colour
  • Self standing, with 1D and 2D barcode
    2,00 ml
    Round bottom
    Yes
    Orange
    1 * 500 ST
    1 * 500 ST - 585,00 €
    585,00 €
    1 * 500 ST
       585,00 €
  • Self standing, with 1D barcode
    2,00 ml
    Round bottom
    Yes
    Orange
    1 * 500 ST
    1 * 500 ST - 386,00 €
    386,00 €
    1 * 500 ST
       386,00 €
  • Self standing, with 1D and 2D barcode
    2,00 ml
    Round bottom
    Yes
    Orange
    1 * 500 ST
    1 * 500 ST - 585,00 €
    585,00 €
    1 * 500 ST
       585,00 €
  • Self standing, with 1D barcode
    2,00 ml
    Round bottom
    Yes
    Orange
    1 * 500 ST
    1 * 500 ST - 386,00 €
    386,00 €
    1 * 500 ST
       386,00 €
Cryogenic storage boxes
  •  
    Colour
    Compartments
    For
  • White
    9×9
    1 and 2 ml tubes
    1 * 10 ST
    1 * 10 ST - 144,00 €
    144,00 €
    1 * 10 ST
       144,00 €
  •  
    Colour
    Compartments
    For
  • Orange
    10×10
    1 and 2 ml tubes
    1 * 10 ST
    1 * 10 ST - 144,00 €
    144,00 €
    1 * 10 ST
       144,00 €