Hanging racks
About this item
Our stainless-steel hanging racks are manufactured to the highest standards.
- Brushed dull polish finish
- Wall mounted
- Optional number etching if required
Our hanging racks are manufactured with 304 grade stainless steel for optimised durability and strength.
Long-lasting and sturdy, they are designed to the highest standard, ensuring super-smooth edges to minimise scratching and snagging.
Wall mounted, enabling space-efficiency. As with all our products, our stainless-steel coat racks are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
Available in a variety of sizes.
8 Options Available Below
Product Details & Documents
Our stainless-steel hanging racks are manufactured to the highest standards.
- Brushed dull polish finish
- Wall mounted
- Optional number etching if required
Our hanging racks are manufactured with 304 grade stainless steel for optimised durability and strength.
Long-lasting and sturdy, they are designed to the highest standard, ensuring super-smooth edges to minimise scratching and snagging.
Wall mounted, enabling space-efficiency. As with all our products, our stainless-steel coat racks are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
Available in a variety of sizes.
Product Family Options
Product Information
- AvailabilityPrice
- TEKMCRGS-HR2HHanging rack, 2 hooks, 165W×95H mm
- TEKMWR0303Hanging rack, 3 hooks, 165W×160H mm
- TEKMWR0206/316Hanging rack, 6 hooks, 730W×95H mm
- TEKMCRGS-HR8HHanging rack, 8 hooks, 1000W×95H mm
- TEKMCRGS-HR12HHanging rack, 12 hooks, 1500W×95H mm
- TEKMWR0315Hanging rack, 15 hooks, 980W×160H mm
- TEKMWR0319Hanging rack, 19 hooks, 1250W×160H mm
- TEKMWR0323Hanging rack, 23 hooks, 1480W×160H mm