Orders LinkContactUsLinkComponent
Four drawer storage cupboards
Four drawer storage cupboards
 :   TEKNOMEK
Four drawer storage cupboards
 :  TEKNOMEK
Four drawer storage cupboards
Our storage cupboard comes complete with four drawers to provide perfect storage.
  • Brushed dull polish finish
  • Sloping top
  • Four shelf height adjustments
  • Stainless steel adjustable feet with non-marking rubber base
  • Lockable main doors
  • Four internal or external drawers
  • Shelves sold separately
Our cupboard has been designed with a sloping top, four drawers (internal and external options available) and lockable doors to secure all content.
The range includes a single (600 mm) or double (1000 or 1200 mm) solid door.
Supplied with four easy clean shelf height adjustments, just add the number of shelves necessary to suit your needs.
Our cupboards are tough, with a robust 1,5 mm thick stainless steel, they are designed with hygiene in mind. No ledges or crevices assures that dirt cannot find anywhere to hide, while the solid stainless steel construction enables easy cleaning. Further, the 300 mm legs allows easy access to any dirt hiding beneath and audit inspections.
Product Family Options
  • Description
    Material
    W×D×H
  • Storage cupboard with external drawers
    Stainless steel 304
    1000×600×2080 mm
    1 * 1 ST
    1 * 1 ST
  • Storage cupboard with internal drawers
    Stainless steel 304
    1000×600×2080 mm
    1 * 1 ST
    1 * 1 ST
  • Storage cupboard with external drawers
    Stainless steel 304
    1200×600×2080 mm
    1 * 1 ST
    1 * 1 ST
  • Storage cupboard with internal drawers
    Stainless steel 304
    1200×600×2080 mm
    1 * 1 ST
    1 * 1 ST