ContactUsLinkComponent
24 clear not treated reservoir, sterile, HTS Transwell®
24 clear not treated reservoir, sterile, HTS Transwell®
Catalog # 734-1558
Supplier:  Corning
   Low Manufacturing Impact
undefined
24 clear not treated reservoir, sterile, HTS Transwell®
Catalog # 734-1558
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • Sterile:
    +
  • Description:
    HTS Transwell® non treated reservoir
  • Seq:
    07
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Pk:
    48
Specifications
About this item
For use as reservoir tray with HTS Transwell permeable supports.
  • Not treated for cell attachment
  • Same footprint as standard 24 well plate but with single rectangular well