ContactUsLinkComponent
Fibronectin 96-well clear flat bottom TC-treated microplates, Corning® BioCoat®
Fibronectin 96-well clear flat bottom TC-treated microplates, Corning® BioCoat®
Catalog # 734-0126
Supplier:  Corning
   Low Manufacturing Impact
undefined
Fibronectin 96-well clear flat bottom TC-treated microplates, Corning® BioCoat®
Catalog # 734-0126
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of wells:
    96
  • Packed:
    5 plates
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    96-well plates
  • Seq:
    03
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 - Bedford
  • Culture area:
    0,32 cm²
  • Pk:
    5