ContactUsLinkComponent
96-Well clear polypropylene PCR microplate, Corning® Thermowell® GOLD
96-Well clear polypropylene PCR microplate, Corning® Thermowell® GOLD
Catalog # 734-4137
Supplier:  Corning
   Low Manufacturing Impact
undefined
96-Well clear polypropylene PCR microplate, Corning® Thermowell® GOLD
Catalog # 734-4137
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of wells:
    96
  • Skirt:
    Semi-skirted
  • Well colour:
    Clear
  • Description:
    PCR plate, with 2 barcode labels, Thermowell® GOLD
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Barcoded:
    Yes
  • Pk:
    50
Specifications
About this item
These half skirted 96 well PCR microplates feature a rigid construction that is ideal for automation.
  • Clear medical grade PP is suitable for RT-PCR
  • Uniform wall thinness assures efficient heat transfer while minimizing well-to-well variability
  • 100% tested for well integrity and DNase- and RNase-free
  • Nonsterile, without lids
  • With two generic bar code labels per plate