ContactUsLinkComponent
Cleanroom overalls, sterile, with attached hood, CleanMax®
Cleanroom overalls, sterile, with attached hood, CleanMax®
Certificates
undefined
Cleanroom overalls, sterile, with attached hood, CleanMax®
Specifications
  • Catalog No:
  • 114-9538
  • 114-9537
  • 114-9539
  • 114-9536
  • 114-9535
  • 114-9541
  • 114-9540
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Cleanroom class:
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom