96-well plates, polypropylene, without lids
About this item
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
9 Options Available Below
Product Details & Documents
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
Ordering information: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
Specifications for Product Family
Specifications
- Catalog No:
- 734-1545
- 734-1550
- 734-1781
- 734-1782
- 734-1547
- 734-1552
- 734-1551
- 732-0484
- 732-0485
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
Product Information
- Well volumeSterilePackedAvailabilityPrice
- 734-1545Storage plate, 96-well, natural, V-shaped wellStorage plate, 96-well, natural, V-shaped well320 µl+25/bagLog in for availability1 * 100 ST404,00 €Specifications:
More
Specifications
Cat. No.734-1545Environmentally PreferableEnvironmental certification - 734-1550Storage plate, 96-well, natural, V-shaped wellStorage plate, 96-well, natural, V-shaped well320 µl-25/bagLog in for availability1 * 100 ST350,00 €Specifications:
More
Specifications
Cat. No.734-1550Environmentally PreferableEnvironmental certification - 734-1781Storage plate, 96-well, white, round wellSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1781Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact - 734-1782Storage plate, 96-well, black, round wellSpecifications:
Low Manufacturing Impact More
Specifications
Cat. No.734-1782Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationRECs, ISO 14001 - KennebunkLow Manufacturing Impact - 734-1547Storage plate, 96-well, natural, round wellSpecifications:
More
Specifications
Cat. No.734-1547Environmentally PreferableEnvironmental certification - 734-1552Storage plate, 96-well, natural, round wellSpecifications:
More
Specifications
Cat. No.734-1552Environmentally PreferableEnvironmental certification - 734-1551Storage plate, 96-well, natural, flat wellSpecifications:
More
Specifications
Cat. No.734-1551Environmentally PreferableEnvironmental certification - 732-0484Storage plate, 96-well, expanded volume, natural, V-bottom wellStorage plate, 96-well, expanded volume, natural, V-bottom well450 µl-10/bagLog in for availability1 * 50 ST327,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.732-0484Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact - 732-0485Storage plate, 96-well, expanded volume, natural, V-bottom wellStorage plate, 96-well, expanded volume, natural, V-bottom well450 µl+10/bagLog in for availability1 * 50 ST373,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.732-0485Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationI-RECs, ISO 14001 - ReynosaLow Manufacturing Impact