Modified Impeller Assembly for Magnetically-Driven Corning® Proculture® Spinner Flask
Supplier: Corning
Certificates
About this item
Stainless steel impeller shaft with dual bearing, modified impeller assembly for use in magnetically driven, Proculture® Spinner Flasks
- Narrower impeller paddle design allows use with probes to create a small bioreactor
- Non sterile
Frequently Bought Together