Order Entry
Belgium
ContactUsLinkComponent
Magbeads FastDNA kit for FFPE, 96 preparations, MP Biomedicals
Magbeads FastDNA kit for FFPE, 96 preparations, MP Biomedicals
Catalog # 116576096.
Supplier:  MP Biomedicals
CAS Number:  
undefined
Magbeads FastDNA kit for FFPE, 96 preparations, MP Biomedicals
Catalog # 116576096.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    DNA
  • Description:
    Magbeads FastDNA kit for FFPE

Specifications

About this item

MagBeads FastDNA® Kit for FFPE is intended for rapid extraction of DNA from FFPE sample. This kit uses two combination methods. High-salt Bind is conducive to remove pigments or polysaccharides from complex FFPE samples, so as to improve the purity of nucleic acid and avoid blocking aligent 2100. Alcohol mediated adsorption is conducive for improving the nucleic acid yield.

  • High quality DNA: intact, high yields and free from inhibitors
  • High throughput: compatible with a wide range of automated nucleic acid purification platforms
  • Flexibility: manual and automation mode of extraction
  • Environmentally friendly: no phenol/chloroform or other toxic chemicals

MagBeads FastDNA® Kit for FFPE is based on the purification method of high binding magnetic particles. The sample is lysed and digested. RNA is released into the lysate. After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.

The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts. Finally the RNA was eluted with elution buffer.