Order Entry
Belgium
ContactUsLinkComponent
Magbeads FastRNA kit for FFPE, 192 preparations, MP Biomedicals
Magbeads FastRNA kit for FFPE, 192 preparations, MP Biomedicals
Catalog # 116573192.
Supplier:  MP Biomedicals
CAS Number:  
undefined
Magbeads FastRNA kit for FFPE, 192 preparations, MP Biomedicals
Catalog # 116573192.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    192 Tests
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    FFPE tissues
  • Target molecule:
    RNA
  • Description:
    Magbeads FastRNA kit for FFPE

Specifications

About this item

MagBeads FastRNA Kit for FFPE is intended for rapid extraction of RNA from tissue, cells, blood, and other clinical samples. RNA can be used directly for RT-PCR, quantitative RT-PCR and so on.

  • High quality RNA: intact, high yields and free from inhibitors
  • High throughput: compatible with a wide range of automated nucleic acid purification platforms
  • Flexibility: manual and automation mode of extraction
  • Environmentally friendly: no phenol/chloroform or other toxic chemicals

MagBeads FastRNA Kit for FFPE is based on the purification method of high binding magnetic particles. The sample is lysed and digested. RNA is released into the lysate. After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption.

The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts. Finally the RNA was eluted with elution buffer.