Magbeads FastRNA kit, 96 preparations, MP Biomedicals
N° de catalogue 116572096.
Fournisseur: MP Biomedicals
Numéro CAS:
Spécifications
- Pk:96 Tests
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Human/animal cells and tissues
- Target molecule:RNA
- Description:Magbeads FastRNA kit
Spécifications
À propos de cet article
MagBeads FastRNA Kit is intended for simple and rapid extraction of total RNA from tissue and culture cells samples.
- High quality RNA: intact, high yields and free from inhibitors
- High throughput: compatible with a wide range of automated nucleic acid purification platforms
- Flexibility: manual and automation mode of extraction
- Environmentally friendly: no phenol/chloroform or other toxic chemicals
MagBeads FastRNA Kit is intended for simple and rapid extraction of total RNA from tissue and culture cells samples and is based on superparamagnetic particles purification technology, no phenol-chloroform extraction or alcohol precipitation. The whole extraction process is only 60 minutes.
Purified RNA is ready for downstream applications such as RTPCR, virus RNA testing and so on. The kits buffers can be used for both manual extraction process and automatic nucleic acid extraction machines.